Specifications
Assay Percent Notes(metals basis)
CAS118448-18-3
Chemical Name or MaterialBis(2,2,6,6-tetramethyl-3,5-heptanedionato)calcium(II)
Formula Weight406.57
IUPAC Namecalcium;(Z)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate;(E)-2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate
View more