Silver powder, spherical, -635 mesh, 99.9% (metals basis)
Silver powder, spherical, -635 mesh, 99.9% (metals basis)
Silver powder, spherical, -635 mesh, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver powder, spherical, -635 mesh, 99.9% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
250 g
Catalog number 041600.09
also known as 041600-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Specifications
CAS7440-22-4
Chemical Name or MaterialSilver powder
DOT InformationHazard Class: 9; Packaging Group: III
EINECS Number231-131-3
IUPAC Namesilver
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only