Cobalt powder, spherical, -100+325 mesh, 99.8% (metals basis)
Cobalt powder, spherical, -100+325 mesh, 99.8% (metals basis)
Cobalt powder, spherical, -100+325 mesh, 99.8% (metals basis)
Thermo Scientific Chemicals

Cobalt powder, spherical, -100+325 mesh, 99.8% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 041605.18
also known as 041605-18
Price (USD)
94.65
Special offer
Online exclusive
Ends: 30-Jun-2026
111.00
Save 16.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
94.65
Special offer
Online exclusive
Ends: 30-Jun-2026
111.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
Appearance (Color)Gray
FormPowder
Comment-100+325 Mesh
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only