Copper(II) methacrylate, tech.
Copper(II) methacrylate, tech.
Copper(II) methacrylate, tech.
Thermo Scientific Chemicals

Copper(II) methacrylate, tech.

CAS: 19662-59-0 | C4H5CuO2 | 148.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 041621.14
also known as 041621-14
Price (USD)
217.65
Special offer
Online exclusive
Ends: 30-Jun-2026
256.00
Save 38.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
217.65
Special offer
Online exclusive
Ends: 30-Jun-2026
256.00
Save 38.35 (15%)
Each
Chemical Identifiers
CAS19662-59-0
IUPAC Namecopper(2+) 2-methylprop-2-enoate
Molecular FormulaC4H5CuO2
InChI KeyBFHDHRZKYIRNRP-UHFFFAOYSA-M
SMILES[Cu++].CC(=C)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to green
Formpowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only