Strontium titanium oxide, S.A. 12.5-25m<sup>2</sup>/g
Strontium titanium oxide, S.A. 12.5-25m<sup>2</sup>/g
Strontium titanium oxide, S.A. 12.5-25m<sup>2</sup>/g
Thermo Scientific Chemicals
Strontium titanium oxide, S.A. 12.5-25m2/g
CAS: 12060-59-2 | O3SrTi | 183.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 041634.36
also known as 041634-36
Price (USD)
725.65
Special offer
Online exclusive
Ends: 30-Jun-2026
854.00
Save 128.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
725.65
Special offer
Online exclusive
Ends: 30-Jun-2026
854.00
Save 128.35 (15%)
Each
Chemical Identifiers
CAS12060-59-2
SpecificationsSpecification SheetSpecification Sheet
Comment12.5-25m²/g
Comment0.05-0.15 Micron APS
Strontium titaniumoxide is used as paint . It has been used as a strontium-90-containing material in radioisotope thermoelectric generators. It is also used as a diamond simulant. This compound is been used in television sets, radios and computers, capacitors, because of the dielectric constant of the material. Used in electronics and in electrical insulation.