Lanthanum(III) oxide, 99.9% (metals basis)
Lanthanum(III) oxide, 99.9% (metals basis)
Lanthanum(III) oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Lanthanum(III) oxide, 99.9% (metals basis)

For vacuum deposition | CAS: 1312-81-8
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 041644.18
also known as 041644-18
Price (USD)
164.65
Special offer
Online exclusive
Ends: 30-Jun-2026
194.00
Save 29.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
164.65
Special offer
Online exclusive
Ends: 30-Jun-2026
194.00
Save 29.35 (15%)
Each
Chemical Identifiers
CAS1312-81-8
IUPAC Namedilanthanum(3+) trioxidandiide
Molecular FormulaLa2O3
InChI KeyMRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[La+3].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Lanthanum(III) oxide is used for vacuum deposition. It is also acts as a precursor to LAMOX fast ion conductors. It is also employed in the manufacture of special alloys, optical glass and refractory materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only