Terbium(III) chloride, anhydrous, 99.9% (REO)
Terbium(III) chloride, anhydrous, 99.9% (REO)
Terbium(III) chloride, anhydrous, 99.9% (REO)
Terbium(III) chloride, anhydrous, 99.9% (REO)
Thermo Scientific Chemicals

Terbium(III) chloride, anhydrous, 99.9% (REO)

CAS: 10042-88-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 041678.06
also known as 041678-06
Price (USD)
285.65
Special offer
Online exclusive
Ends: 31-Dec-2025
336.00
Save 50.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
285.65
Special offer
Online exclusive
Ends: 31-Dec-2025
336.00
Save 50.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS10042-88-3
IUPAC Nametrichloroterbium
Molecular FormulaCl3Tb
InChI KeyGFISHBQNVWAVFU-UHFFFAOYSA-K
SMILESCl[Tb](Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only