Multi Ion anion IC standard solution in H{2}O, Specpure™
Thermo Scientific Chemicals

Multi Ion anion IC standard solution in H{2}O, Specpure™

 
Have Questions?
Catalog NumberQuantity
041694.AE
also known as 041694-AE
100 mL
Catalog number 041694.AE
also known as 041694-AE
Price (USD)
230.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
230.00
Each
Specifications
Chemical Name or MaterialMulti Ion anion IC standard solution in H2O
EINECS Number231-791-2
MDL NumberMFCD00144813
Physical FormLiquid
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only