Specifications
Chemical Name or MaterialSulfur in Diesel Fuel standard solution
EINECS Number270-676-1
Health Hazard 1H315-H319-H335-H351
Health Hazard 2GHS H Statement
H351-H315-H319-H335-H227
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
View more