Silicon(II) oxide, 99.9% (metals basis)
Silicon(II) oxide, 99.9% (metals basis)
Silicon(II) oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Silicon(II) oxide, 99.9% (metals basis)

CAS: 10097-28-6 | OSi | 44.084 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 041710.36
also known as 041710-36
Price (USD)
719.65
Special offer
Online exclusive
Ends: 30-Jun-2026
847.00
Save 127.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
719.65
Special offer
Online exclusive
Ends: 30-Jun-2026
847.00
Save 127.35 (15%)
Each
Chemical Identifiers
CAS10097-28-6
IUPAC Nameoxidaniumylidynesilanide
Molecular FormulaOSi
InChI KeyLIVNPJMFVYWSIS-UHFFFAOYSA-N
SMILES[O+]#[Si-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only