Sodium diphenylamine-4-sulfonate, ACS
Sodium diphenylamine-4-sulfonate, ACS
Sodium diphenylamine-4-sulfonate, ACS
Thermo Scientific Chemicals

Sodium diphenylamine-4-sulfonate, ACS

Redox indicator | CAS: 6152-67-6 | C12H11NNaO3S | 272.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 041714.09
also known as 041714-09
Price (USD)
97.80
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
97.80
Each
Chemical Identifiers
CAS6152-67-6
IUPAC Name4-(phenylamino)benzene-1-sulfonic acid sodium
Molecular FormulaC12H11NNaO3S
InChI KeyVMVSDVIYIMNZBS-UHFFFAOYSA-N
SMILES[Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to grey
CommentSensitivity as indicator P.T.
Formpowder
Redox indicatorSodium diphenylamine-4-sulfonate is used as a pH independent redox indicator used in the determination of redox potential. It is used in the preparation of poly-(aninile-co-N-(4-sulfophenyl)aniline) by polymerization with aniline. It acts as a ligand and form complexes with metal halides like lanthanum(III) chloride. It is also involved in the synthesis of gold nano crystals by reaction with chloroauric acid in the presence of polyethylene glycol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only