Potassium aquapentachlororuthenate(III), Premion™, 99.99% (metals basis), Ru 26.4% min
Potassium aquapentachlororuthenate(III), Premion™, 99.99% (metals basis), Ru 26.4% min
Potassium aquapentachlororuthenate(III), Premion™, 99.99% (metals basis), Ru 26.4% min
Thermo Scientific Chemicals

Potassium aquapentachlororuthenate(III), Premion™, 99.99% (metals basis), Ru 26.4% min

CAS: 14404-33-2 | Cl5K2Ru | 356.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 041716.03
also known as 041716-03
Price (USD)
82.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.50
Save 14.85
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
82.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.50
Save 14.85
Each
Chemical Identifiers
CAS14404-33-2
IUPAC Namedipotassium pentachlororutheniumdiuide
Molecular FormulaCl5K2Ru
InChI KeyIXYMQZXAJLWXRK-UHFFFAOYSA-I
SMILES[K+].[K+].Cl[Ru--](Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Potassium aquapentachlororuthenate(III) is used in Hydrogenation (Slurry Bed).It is also used as an intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only