Cadmium nitrate tetrahydrate, 99.9% (metals basis)
Cadmium nitrate tetrahydrate, 99.9% (metals basis)
Cadmium nitrate tetrahydrate, 99.9% (metals basis)
Thermo Scientific Chemicals

Cadmium nitrate tetrahydrate, 99.9% (metals basis)

CAS: 10022-68-1 | CdH2N2O6 | 238.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041732.30
also known as 041732-30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS10022-68-1
IUPAC Namecadmium(2+) bis(nitric acid)
Molecular FormulaCdH2N2O6
InChI KeyIZIPRBSCGMISKX-UHFFFAOYSA-N
SMILES[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Cadmium nitrate tetrahydrate is used in Photographic emulsions, coloring glass and porcelain, laboratory reagent, and the production of other cadmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only