Humic acid
Thermo Scientific Chemicals

Humic acid

CAS: 1415-93-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 041747.14
also known as 041747-14
Price (USD)
168.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
168.00
Each
Chemical Identifiers
CAS1415-93-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark brown to black
FormCrystalline powder or powder or crystals
Assay from Suppliers CofA≥95% (TLC)
Metal chelator; pigments.Humic acid is used as a soil supplement in agriculture and human nutritional supplement. It is used to improve the growth and cultivation of crops, citrus, turf, flowers. It is also used to improve the strength of organically-deficient soils. It is utilized to stimulate the immune system and treating the influenza, avian flu, swine flu and other viral infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only