Humic acid sodium salt, tech. 50-60% (as humic acid)
Humic acid sodium salt, tech. 50-60% (as humic acid)
Humic acid sodium salt, tech. 50-60% (as humic acid)
Thermo Scientific Chemicals

Humic acid sodium salt, tech. 50-60% (as humic acid)

CAS: 68131-04-4 | C9H8Na2O4 | 226.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 041748.36
also known as 041748-36
Price (USD)
155.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
155.00
Each
Chemical Identifiers
CAS68131-04-4
IUPAC Namedisodium bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylate
Molecular FormulaC9H8Na2O4
InChI KeyHMGUIQPKFUZDPV-UHFFFAOYNA-L
SMILES[Na+].[Na+].[O-]C(=O)C1C2CC(C=C2)C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)45 to 70% ( as Humic acid)
pH9-10
Loss on Drying15% max
Humic acid sodium salt is a principal component of humic substances. It boosts plant growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only