Specifications
Chemical Name or MaterialSulfur in Residual Oil standard solution
Health Hazard 1H335-H350
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P201-P202-P281-P308+P313-P501c
MDL NumberMFCD00197838
View more