Sulfur in Residual Oil standard solution, Specpure™, 30,000μg/g (3.00%)
Thermo Scientific Chemicals

Sulfur in Residual Oil standard solution, Specpure™, 30,000μg/g (3.00%)

 
Have Questions?
Catalog NumberQuantity
041756.AE
also known as 041756-AE
100 mL
Catalog number 041756.AE
also known as 041756-AE
Price (USD)
202.65
Special offer
Online exclusive
Ends: 30-Jun-2026
238.00
Save 35.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
202.65
Special offer
Online exclusive
Ends: 30-Jun-2026
238.00
Save 35.35 (15%)
Each
Specifications
Chemical Name or MaterialSulfur in Residual Oil standard solution
Health Hazard 1H315-H319-H335-H350
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
MDL NumberMFCD00197838
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only