Sodium oxalate, ACS, 99.5+%
Sodium oxalate, ACS, 99.5+%
Sodium oxalate, ACS, 99.5+%
Thermo Scientific Chemicals

Sodium oxalate, ACS, 99.5+%

CAS: 62-76-0 | C2Na2O4 | 134.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 041759.A1
also known as 041759-A1
Price (USD)
242.00
Each
Quantity:
1 kg
Request bulk or custom format
Price (USD)
242.00
Each
Chemical Identifiers
CAS62-76-0
IUPAC Namedisodium oxalate
Molecular FormulaC2Na2O4
InChI KeyZNCPFRVNHGOPAG-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentK 0.005%
Impurity contentNH₄ 0.002%
Impurity contentSO₄ 0.002%
Impurity contentSubstances darkened by hot sulfuric acid P.T.
Insoluble material0.005%
View more
Sodium oxalate is used as a reducing agent, as a primary standard for standardizing potassium permanganate solutions and as a neutralizer for exothermic acidic reactions. It finds application in metal cleaning preparations, metal extraction and separation, blueprint coatings, electroplating baths, special cement manufacturing, plant nutrition as well as in textile and pharmaceutical industries. It plays an important role to remove calcium ions from blood plasma and prevents blood clotting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only