Zinc acrylate
Zinc acrylate
Zinc acrylate
Thermo Scientific Chemicals

Zinc acrylate

CAS: 14643-87-9 | C6H6O4Zn | 207.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 041780.18
also known as 041780-18
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.00
Save 6.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
35.65
Special offer
Online exclusive
Ends: 30-Jun-2026
42.00
Save 6.35 (15%)
Each
Chemical Identifiers
CAS14643-87-9
IUPAC Namezinc(2+) bis(prop-2-enoate)
Molecular FormulaC6H6O4Zn
InChI KeyXKMZOFXGLBYJLS-UHFFFAOYSA-L
SMILES[Zn++].[O-]C(=O)C=C.[O-]C(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay from Supplier's CofA≥94%
Melting Point> 250?C
The primary use of Zinc Acrylate is in the production of golf balls. Other uses are primarily in the production of specialty rubber products such as belts and hoses for automobiles. Also used as a tissue adhesive used for adherence of tissue to tissue or tissue to non-tissue for prostheses in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only