3,4-Difluorotoluene, 98%
3,4-Difluorotoluene, 98%
3,4-Difluorotoluene, 98%
3,4-Difluorotoluene, 98%
Thermo Scientific Chemicals

3,4-Difluorotoluene, 98%

CAS: 2927-34-6 | C7H6F2 | 128.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 041789.03
also known as 041789-03
Price (USD)
34.90
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.90
Each
Add to cart
Specifications
CAS2927-34-6
Chemical Name or Material3,4-Difluorotoluene
DOT InformationTransport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density1.120 g/mL
Flash Point25°C (77°F)
View more
Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only