Aluminum Thinfoil, 10.0 micron (0.0004in) thick, 99.0% (metals basis)
Aluminum Thinfoil, 10.0 micron (0.0004in) thick, 99.0% (metals basis)
Aluminum Thinfoil, 10.0 micron (0.0004in) thick, 99.0% (metals basis)
Thermo Scientific Chemicals

Aluminum Thinfoil, 10.0 micron (0.0004in) thick, 99.0% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
Catalog number 041798.FI
also known as 041798-FI
Price (USD)
48.65
Online exclusive
54.10
Save 5.45
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
48.65
Online exclusive
54.10
Save 5.45
Each
Chemical Identifiers
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
Assay from Supplier's CofA≥99.0% (metals basis)
FormFoil, 10.0 micron (0.0004in) thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only