Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 041804.FF
also known as 041804-FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangePt:Ir; 90:10 wt%
Chemical Name or MaterialPlatinum Iridium foil
MDL NumberMFCD02091683
Name Note0.1mm (0.004 in.) thick
View more

Platinum Iridium foil, 0.1mm (0.004in) thick, 99.9% (metals basis) is used in non-standard thermocouples, electrical contacts and as electrodes in medical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only