Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 041805.FF
also known as 041805-FF
Price (USD)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangePt:Ir; 80:20 wt%
Chemical Name or MaterialPlatinum Iridium foil
MDL NumberMFCD02091683
Name Note0.025mm (0.001 in.) thick
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only