Palladium, 5% on 3mm alumina pellets
Palladium, 5% on 3mm alumina pellets
Palladium, 5% on 3mm alumina pellets
Thermo Scientific Chemicals

Palladium, 5% on 3mm alumina pellets

Hydrogen removal 'getter' | CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Catalog NumberQuantity
041825.06
also known as 041825-06
5 g
Catalog number 041825.06
also known as 041825-06
Price (USD)
79.65
Online exclusive
88.40
Save 8.75
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
79.65
Online exclusive
88.40
Save 8.75
Each
Specifications
CAS5-3-7440
ChEBICHEBI:33363
Chemical Name or MaterialPalladium
IUPAC Namepalladium
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
View more
Used as hydrogen removal getter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only