Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles
Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles
Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Methanol, anhydrous, 99.9%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 041838.K2
also known as 041838-K2
Price (USD)
63.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
63.00
Each
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>/= 99.9%
Appearance (Color)Clear, colorless
FormLiquid
Water Content (Karl Fischer Titration)100 ppm, max
Residue on Evaporation3 ppm, max
Methanol is widely used as a solvent in synthetic organic chemistry. It is also used as a solvent in analytical chemistry, especially for HPLC, LC-MS, GC-MS, ICP-MS and spectrophotometry. It is used in the production of biodiesel, formaldehyde and acetic acid. It is also used as fuel, antifreeze, denaturant for ethanol and an energy carrier for fuel cell application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only