(1-Hexadecyl)trimethylammonium chloride, 96%
(1-Hexadecyl)trimethylammonium chloride, 96%
(1-Hexadecyl)trimethylammonium chloride, 96%
Thermo Scientific Chemicals

(1-Hexadecyl)trimethylammonium chloride, 96%

CAS: 112-02-7 | C19H42ClN | 320.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 041861.06
also known as 041861-06
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.10
Save 6.45 (16%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 30-Jun-2026
41.10
Save 6.45 (16%)
Each
Chemical Identifiers
CAS112-02-7
IUPAC Namehexadecyltrimethylazanium chloride
Molecular FormulaC19H42ClN
InChI KeyWOWHHFRSBJGXCM-UHFFFAOYSA-M
SMILES[Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)> 95.0%
Appearance (Color)White to off-white
Formpowder
(1-Hexadecyl)trimethylammonium chloride is used as a topical antiseptic and surfactant. It is used in hair care formulations, cream rinse ingredient, hair conditioners and shampoos as a conditioning agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only