Iron(III) ethoxide, 99.6% (metals basis)
Iron(III) ethoxide, 99.6% (metals basis)
Iron(III) ethoxide, 99.6% (metals basis)
Thermo Scientific Chemicals

Iron(III) ethoxide, 99.6% (metals basis)

Precursor for Sol-Gel processes | CAS: 5058-42-4 | C6H15FeO3 | 191.028 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
10 g
50 g
Catalog number 041863.04
also known as 041863-04
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.60
Save 13.95 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
77.65
Special offer
Online exclusive
Ends: 30-Jun-2026
91.60
Save 13.95 (15%)
Each
Chemical Identifiers
CAS5058-42-4
IUPAC Nameiron(3+) triethanolate
Molecular FormulaC6H15FeO3
InChI KeyJTPUGUWXHGEEHW-UHFFFAOYSA-N
SMILES[Fe+3].CC[O-].CC[O-].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.4% max.
Precursor for Sol-Gel processes.Iron(III) ethoxide is used as a precursor for inorganic compound iron oxide. It is used in the preparation of iron-platinum nanoparticles by pyrolysis with platinum(II) acetylacetonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only