Copper(II) hexafluoro-2,4-pentanedionate hydrate, 99.99% (metals basis)
Copper(II) hexafluoro-2,4-pentanedionate hydrate, 99.99% (metals basis)
Copper(II) hexafluoro-2,4-pentanedionate hydrate, 99.99% (metals basis)
Thermo Scientific Chemicals

Copper(II) hexafluoro-2,4-pentanedionate hydrate, 99.99% (metals basis)

CAS: 155640-85-0 | C10H2CuF12O4 | 477.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 041870.09
also known as 041870-09
Price (USD)
304.65
Special offer
Online exclusive
Ends: 30-Jun-2026
358.00
Save 53.35 (15%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
304.65
Special offer
Online exclusive
Ends: 30-Jun-2026
358.00
Save 53.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
Boiling Point220°C (decomposition)
CAS155640-85-0
Chemical Name or MaterialCopper(II) hexafluoro-2,4-pentanedionate hydrate
EINECS Number238-850-1
View more
Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only