Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Polyimide film, 0.008mm thick, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyimide film, 0.008mm thick, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
150 x 150 mm
300 x 300 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041896.GT
Price (USD)
-
Quantity:
150 x 150 mm
Request bulk or custom format
Specifications
CAS62929-02-6
Chemical Name or MaterialPolyimide film
IUPAC Name1-(4-aminophenyl)-1,3,3-trimethyl-2H-inden-5-amine;5-(1,3-dioxo-2-benzofuran-5-carbonyl)-2-benzofuran-1,3-dione
InChI KeyCDTDIQLZIBORMV-UHFFFAOYNA-N
MDL NumberMFCD00677744
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only