Strontium tungsten oxide, 99.9% (metals basis)
Strontium tungsten oxide, 99.9% (metals basis)
Strontium tungsten oxide, 99.9% (metals basis)
Thermo Scientific Chemicals

Strontium tungsten oxide, 99.9% (metals basis)

CAS: 13451-05-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 041899.22
also known as 041899-22
Price (USD)
425.65
Special offer
Online exclusive
Ends: 30-Jun-2026
501.00
Save 75.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
425.65
Special offer
Online exclusive
Ends: 30-Jun-2026
501.00
Save 75.35 (15%)
Each
Chemical Identifiers
CAS13451-05-3
Molecular FormulaO4SrW
InChI KeyKXELQHMRKFESFN-UHFFFAOYSA-N
SMILES[Sr++].[O-][W]([O-])(=O)=O
Molecular Weight (g/mol)335.46
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only