Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis), Thermo Scientific Chemicals
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis), Thermo Scientific Chemicals
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis), Thermo Scientific Chemicals
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis), Thermo Scientific Chemicals
Thermo Scientific™

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV), 99.7% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
041909.06
also known as 041909-06
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041909.06
also known as 041909-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS63370-90-1
Chemical Name or MaterialTetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV)
Formula Weight910.5
InChI KeyGPCQYWUYUFMJFR-UHFFFAOYSA-N
View more
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)hafnium(IV) is used as an intermediate to fabricate capacitor devices on the nano scale.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only