Polystyrene standard, M.W. 13,000, Mw/Mn 1.06
Polystyrene standard, M.W. 13,000, Mw/Mn 1.06
Polystyrene standard, M.W. 13,000, Mw/Mn 1.06
Thermo Scientific Chemicals

Polystyrene standard, M.W. 13,000, Mw/Mn 1.06

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
041928.03
also known as 041928-03
1 g
Catalog number 041928.03
also known as 041928-03
Price (USD)
85.70
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
85.70
Each
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
Molecular Weight13,000, Mw/Mn 1.06
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only