Polystyrene standard, M.W. 290,000, Mw/Mn 1.06
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06
Polystyrene standard, M.W. 290,000, Mw/Mn 1.06
Thermo Scientific Chemicals

Polystyrene standard, M.W. 290,000, Mw/Mn 1.06

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Catalog NumberQuantity
041937.03
also known as 041937-03
1 g
Catalog number 041937.03
also known as 041937-03
Price (USD)
72.65
Online Exclusive
81.10
Save 8.45 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
81.10
Save 8.45 (10%)
Each
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
Molecular Weight288,000 to 294,000
Appearance (Color)White
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only