Spectroflux 100, low phosphorus, Lithium tetraborate, phosphorus <20ppm
Thermo Scientific Chemicals

Spectroflux 100, low phosphorus, Lithium tetraborate, phosphorus <20ppm

A general purpose flux with a low phosphorus content. Dissolves most refractories. Often used in testing of ceramics. Not suitable for highly acidic samples. Samples must be fully oxidized | CAS: 12007-60-2
Have Questions?
Catalog NumberQuantity
041951.A1
also known as 041951-A1
1 kg
Catalog number 041951.A1
also known as 041951-A1
Price (USD)
418.65
Online Exclusive
465.00
Save 46.35 (10%)
Each
Quantity:
1 kg
Price (USD)
418.65
Online Exclusive
465.00
Save 46.35 (10%)
Each
A general purpose flux with a low phosphorus content. Dissolves most refractories. Often used in testing of ceramics. Not suitable for highly acidic samples. Samples must be fully oxidizedSpectroflux 100, low phosphorus, Lithium tetraborate is used to dissolve refractories. It is a spectroscopic fluxing agent which is utilized for fusing basic oxides, glass, alumino-silicates, ceramics, carbonates, cement, steel, aluminum, soil, concrete mixes, bauxites, magnesites and rare earth oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A general purpose flux with a low phosphorus content. Dissolves most refractories. Often used in testing of ceramics. Not suitable for highly acidic samples. Samples must be fully oxidized
Specifications
CAS12007-60-2
Melting Point920°C
Physical FormSolid
Quantity1 kg
Unit SizeEach