Tricyclohexylphosphine, 98%
Tricyclohexylphosphine, 98%
Tricyclohexylphosphine, 98%
Thermo Scientific Chemicals

Tricyclohexylphosphine, 98%

CAS: 2622-14-2 | C18H33P | 280.436 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 041952.06
also known as 041952-06
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS2622-14-2
IUPAC Nametricyclohexylphosphane
Molecular FormulaC18H33P
InChI KeyWLPUWLXVBWGYMZ-UHFFFAOYSA-N
SMILESC1CCC(CC1)P(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Phosphorus-31 NMR> 97.5%
Used as an active pharmaceutical ingredient. Ligand used in the Pd-catalyzed coupling of malononitrile with aryl halides. Bulky phosphine ligand used with a Pd(0)-triolefinic macrocycle catalyst for Suzuki coupling of aryl bromides and chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only