Iodine, resublimed crystals, ACS, 99.8%
Iodine, resublimed crystals, ACS, 99.8%
Iodine, resublimed crystals, ACS, 99.8%
Thermo Scientific Chemicals

Iodine, resublimed crystals, ACS, 99.8%

CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
200 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041955.36
Price (USD)
292.65
Online Exclusive
325.00
Save 32.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
292.65
Online Exclusive
325.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentChlorine and bromine (as Cl) 0.005%
Impurity contentNonvolatile matter 0.01%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only