Magnesium acrylate
Magnesium acrylate
Magnesium acrylate
Thermo Scientific Chemicals

Magnesium acrylate

CAS: 5698-98-6 | C6H6MgO4 | 166.415 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042002.06
also known as 042002-06
Price (USD)
39.70
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
39.70
Each
Specifications
CAS5698-98-6
Chemical Name or MaterialMagnesium acrylate
EINECS Number227-177-9
Formula Weight166.42
Health Hazard 1Warning
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only