1,18-Octadecanedicarboxylic acid
1,18-Octadecanedicarboxylic acid
1,18-Octadecanedicarboxylic acid
Thermo Scientific Chemicals

1,18-Octadecanedicarboxylic acid

CAS: 2424-92-2 | C20H38O4 | 342.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
250 g
Catalog number 042004.04
also known as 042004-04
Price (USD)
46.60
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
46.60
Each
Specifications
CAS2424-92-2
ChEBICHEBI:73728
Chemical Name or Material1,18-Octadecanedicarboxylic acid
Formula Weight342.52
IUPAC Nameicosanedioic acid
View more
1,18-Octadecanedicarboxylic acid is used as an intermediate in organic synthesis, as dispersing agent, emulsifying agent, as lubricant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only