Dichloromethane, Environmental Grade, 99.8+%, stab. with amylene
Dichloromethane, Environmental Grade, 99.8+%, stab. with amylene
Dichloromethane, Environmental Grade, 99.8+%, stab. with amylene
Thermo Scientific Chemicals

Dichloromethane, Environmental Grade, 99.8+%, stab. with amylene

Suitable For Pesticide Residue Analysis | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042006.K7
also known as 042006-K7
Price (USD)
-
Quantity:
4 L
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)10, max
FormClear liquid
Assay (GC)≥99.8%
Impurity content10 ppt, max- GC-ECD responsive residue ( single peak) as heptachlorepoxide
Impurity contentCholide: 10ppm, max
View more
Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Cl 10ppm, Halogenated residue 10ng/L heptachlorepoxide, Evaporation residue 3ppm, H2O 0.02%
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only