Fullerene powder, sublimed, 99.9+% C{60}
Fullerene powder, sublimed, 99.9+% C{60}
Fullerene powder, sublimed, 99.9+% C{60}
Thermo Scientific Chemicals

Fullerene powder, sublimed, 99.9+% C{60}

CAS: 99685-96-8 | C60 | 720.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
1 g
Catalog number 042008.MF
also known as 042008-MF
Price (USD)
-
Quantity:
50 mg
Request bulk or custom format
Chemical Identifiers
CAS99685-96-8
IUPAC Name(C60-Ih)[5,6]fullerene
Molecular FormulaC60
InChI KeyXMWRBQBLMFGWIX-UHFFFAOYSA-N
SMILESC12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark grey to black
FormCrystalline powder
Assay (HPLC)≥99.9%
Infrared spectrumConforms
The optical absorption properties of C60 match solar spectrum in a way that suggests that C60-based films could be useful for photovoltaic applications. Because of its high electronic affinity it is one of the most common electron acceptor used in donor/acceptor based solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only