Stainless Steel gauze, 150 mesh woven from 0.066mm (0.0026in) dia wire, Type 304
Thermo Scientific Chemicals

Stainless Steel gauze, 150 mesh woven from 0.066mm (0.0026in) dia wire, Type 304

Have Questions?
Change viewbuttonViewtableView
Quantity:
30 x 30 cm
30 x 60 cm
Catalog number 042010.RF
also known as 042010-RF
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Quantity:
30 x 30 cm
Request bulk or custom format
Price (USD)
144.65
Online Exclusive
161.00
Save 16.35 (10%)
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni; 70:19:11 wt%
Chemical Name or MaterialStainless Steel gauze, Type 304
MDL NumberMFCD00148499
Mesh Size150 mesh
Name NoteWoven from 0.066mm (0.0026 in.) dia. wire
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only