2-Hydroxyacetophenone, 97+%
2-Hydroxyacetophenone, 97+%
2-Hydroxyacetophenone, 97+%
Thermo Scientific Chemicals

2-Hydroxyacetophenone, 97+%

CAS: 582-24-1 | C8H8O2 | 136.15 g/mol
Have Questions?
Catalog NumberQuantity
042013.06
also known as 042013-06
5 g
Catalog number 042013.06
also known as 042013-06
Price (USD)
76.65
Online exclusive
84.80
Save 8.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
76.65
Online exclusive
84.80
Save 8.15
Each
Chemical Identifiers
CAS582-24-1
IUPAC Name2-hydroxy-1-phenylethan-1-one
Molecular FormulaC8H8O2
InChI KeyZWVHTXAYIKBMEE-UHFFFAOYSA-N
SMILESOCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)> 97.0%
2-Hydroxyacetophenone can be used to produce 2-hydroxy-1-phenyl-ethanone-(1)-oxime by heating. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only