Stainless Steel spheres, 1mm (0.04in) dia, Type 316
Thermo Scientific Chemicals

Stainless Steel spheres, 1mm (0.04in) dia, Type 316

Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 Pc.
25 Pc.
Catalog number 042018.KN
also known as 042018-KN
Price (USD)
83.00
Each
Quantity:
100 Pc.
Request bulk or custom format
Price (USD)
83.00
Each
Specifications
AppearanceSpheres
Chemical Name or MaterialStainless Steel
Health Hazard 1H317-H351: May cause an allergic skin reaction. Suspected of causing cancer.
MDL NumberMFCD00148499
Molecular FormulaFe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only