Chromium(III) chloride hexahydrate, 98%
Chromium(III) chloride hexahydrate, 98%
Chromium(III) chloride hexahydrate, 98%
Thermo Scientific Chemicals

Chromium(III) chloride hexahydrate, 98%

CAS: 10060-12-5 | Cl3CrH12O6 | 266.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 042113.30
also known as 042113-30
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.00
Save 7.35 (14%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
51.00
Save 7.35 (14%)
Each
Chemical Identifiers
CAS10060-12-5
IUPAC Namechromium(3+) hexahydrate trichloride
Molecular FormulaCl3CrH12O6
InChI KeyLJAOOBNHPFKCDR-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder
Assay from Supplier's CofA≥97.0%
Appearance (Color)Dark green
Impurity content≤0.01% Iron (Fe)
Impurity content≤0.01% Sulfate (SO₄)
View more
Chromium(III) chloride hexahydrate is used as a catalyst, as a mordant in printing and dyeing industry. It is involved in the preparation of anhydrous tetrahydrofuran complex of chromium(III) chloride by reacting with trimethylsilylchloride. It finds application in the manufacturing of a variety of chromium-containing pigment in Paint industry and other chromium salts in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only