Sulfur in Light Mineral Oil standard solution, Specpure™, 20,000μg/g (2.00%)
Thermo Scientific Chemicals

Sulfur in Light Mineral Oil standard solution, Specpure™, 20,000μg/g (2.00%)

 
Have Questions?
Catalog NumberQuantity
042171.AE
also known as 042171-AE
100 mL
Catalog number 042171.AE
also known as 042171-AE
Price (USD)
103.65
Online Exclusive
115.00
Save 11.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
103.65
Online Exclusive
115.00
Save 11.35 (10%)
Each
Specifications
Chemical Name or MaterialSulfur in Light Mineral Oil standard solution
Health Hazard 1H335
Health Hazard 2GHS H Statement
H350-H315-H319
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
P280-P281-P305+P351+P338-P362-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL NumberMFCD00804270
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only