Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper foil, 0.1mm (0.004in) thick, Puratronic™, 99.999% (metals basis)

Form: approx. 2.2g/50x50mm | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
100 x 300 mm
Catalog number 042189.GH
also known as 042189-GH
Price (USD)
267.65
Online Exclusive
297.00
Save 29.35 (10%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
267.65
Online Exclusive
297.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Assay from Supplier's CofA99.999% (metals basis)
FormFoil, 0.1mm (0.004in) thick.
Assay from Supplier's CofA≥99.999% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only