Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis)
Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis)
Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis)
Thermo Scientific Chemicals

Vanadium slug, 11mm (0.43in) dia x 4mm (0.16in) thick, 99.8% (metals basis)

CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 Pc.
25 Pc.
Catalog number 042190.KJ
also known as 042190-KJ
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Quantity:
5 Pc.
Request bulk or custom format
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
Thickness4mm (0.16in) thick.
Assay from Supplier's CofA≥99.8% (metals basis)
FormSlug, 11mm (0.43in) diameter.
Appearance (Color)White silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only