Nickel Chromium wire, 0.25mm (0.01in) dia
Nickel Chromium wire, 0.25mm (0.01in) dia
Nickel Chromium wire, 0.25mm (0.01in) dia
Thermo Scientific Chemicals

Nickel Chromium wire, 0.25mm (0.01in) dia

CAS: 11106-97-1 | CrNi | 110.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 m
100 m
Catalog number 042195.G9
also known as 042195-G9
Price (USD)
43.30
Each
Quantity:
25 m
Request bulk or custom format
Price (USD)
43.30
Each
Chemical Identifiers
CAS11106-97-1
IUPAC Namechromium nickel
Molecular FormulaCrNi
InChI KeyVNNRSPGTAMTISX-UHFFFAOYSA-N
SMILES[Cr].[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.25 mm diameter
Assay from Suppliers CofACr: Report Value (%)
Assay from Suppliers CofANi: ≥77.0%
Impurity contentReport Si, Mn, C, S, P, Pb, Fe %
Nichrome alloys are typically used in resistance wire. They are also used in some dental restorations (fillings) and in other applications. Nichrome is used in a very wide variety of devices where electric heating is required.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only