Silver perchlorate, anhydrous
Silver perchlorate, anhydrous
Silver perchlorate, anhydrous
Silver perchlorate, anhydrous
Thermo Scientific Chemicals

Silver perchlorate, anhydrous

Light Sensitive & Hygroscopic | CAS: 7783-93-9 | AgClO4 | 207.314 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 042209.06
also known as 042209-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7783-93-9
IUPAC Namesilver(1+) perchlorate
Molecular FormulaAgClO4
InChI KeyYDHABVNRCBNRNZ-UHFFFAOYSA-M
SMILES[Ag+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97% min
Silver perchlorate is used as a catalyst in organic chemistry. It is an effective reagent for replacing halides ligands with perchlorate. It is also used in the manufacturing of explosives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only