Indium(III) acetate, 99.99% (metals basis)
Indium(III) acetate, 99.99% (metals basis)
Indium(III) acetate, 99.99% (metals basis)
Thermo Scientific Chemicals

Indium(III) acetate, 99.99% (metals basis)

CAS: 25114-58-3 | C2H4InO2 | 174.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 042230.18
also known as 042230-18
Price (USD)
372.65
Online Exclusive
414.00
Save 41.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
372.65
Online Exclusive
414.00
Save 41.35 (10%)
Each
Chemical Identifiers
CAS25114-58-3
IUPAC Nameindium(3+) triacetate
Molecular FormulaC6H9InO6
InChI KeyVBXWCGWXDOBUQZ-UHFFFAOYSA-K
SMILES[In+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Indium(III) acetate is used as a catalyst in intermolecular radical addition of organic iodides to electron deficient alkenes. It is also used in the reduction of 1-bromo-3-phenylpropane to propylbenzene. Further, it is used to prepare the indium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only